siRNA Details
Gene Symbol: LINC01444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110783.1 | 2 | 678 | |||
GenBank mRNA | CF995332.1 | - | 2 | 496 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000580867.1 | - | 2 | 678 | Chr. 18 - 14969606 - 14970468 [-] on Build GRCh38 |
