siRNA Details
Gene Symbol: LINC00864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046091.1 | 3 | 1101 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000582543.2 | - | 1102 | Chr. 10 - 46740596 - 46751241 [+] on Build GRCh38 |
