siRNA Details
| Gene Symbol: MAPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024559.1 | 2 | 196 | |||
| GenBank mRNA | AW770488.1 | - | 2 | 196 | ||
| BC018035.1 | - | 2 | 324 | |||
| BM714794.1 | - | 1 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAPT-AS1| Gene Symbol: MAPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024559.1 | 2 | 196 | |||
| GenBank mRNA | AW770488.1 | - | 2 | 196 | ||
| BC018035.1 | - | 2 | 324 | |||
| BM714794.1 | - | 1 | 102 | |||