siRNA Details
| Gene Symbol: LOC101928251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135635.1 | 2 | 1382 | |||
| GenBank mRNA | BC041474.1 | - | 2 | 1380 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000583547.2 | - | 2 | 1453 | Chr. 17 - 73513685 - 73518770 [+] on Build GRCh38 | |

LOC101928251
Genomic Map