siRNA Details
| Gene Symbol: LOC101927877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135634.1 | 1 | 256 | |||
| GenBank mRNA | BC040294.1 | - | 1 | 258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927877| Gene Symbol: LOC101927877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135634.1 | 1 | 256 | |||
| GenBank mRNA | BC040294.1 | - | 1 | 258 | ||