siRNA Details
| Gene Symbol: MIR133A1HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110369.1 | 3 | 2428 | |||
| GenBank mRNA | BX648566.1 | - | 3 | 2429 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000581072.5 | - | 3 | 2428 | Chr. 18 - 21825487 - 21831406 [-] on Build GRCh38 | |

MIR133A1HG
Genomic Map