siRNA Details
Gene Symbol: PCAT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024259.1 | 2 | 2070 | |||
GenBank mRNA | AK056805.1 | - | 2 | 2070 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000579458.1 | - | 2 | 2070 | Chr. 18 - 26687621 - 26703638 [-] on Build GRCh38 |
