siRNA Details
Gene Symbol: LOC101929511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135639.1 | 2 | 1580 | |||
GenBank mRNA | BC033560.1 | - | 2 | 1580 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000581489.1 | - | 2 | 1580 | Chr. 17 - 82290046 - 82292814 [-] on Build GRCh38 |
