siRNA Details
Gene Symbol: LOC101927989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136504.1 | 2 | 502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000581996.2 | - | 2 | 502 | Chr. 18 - 76491652 - 76493531 [+] on Build GRCh38 |
