siRNA Details
| Gene Symbol: LOC101927989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136504.1 | 2 | 849 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000581996.2 | - | 849 | Chr. 18 - 76491652 - 76493531 [+] on Build GRCh38 | ||

LOC101927989
Genomic Map