siRNA Details
| Gene Symbol: LOC101927989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136504.1 | 2 | 144 | |||
| GenBank mRNA | BG329748.1 | - | 2 | 144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927989| Gene Symbol: LOC101927989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136504.1 | 2 | 144 | |||
| GenBank mRNA | BG329748.1 | - | 2 | 144 | ||