siRNA Details
| Gene Symbol: CYP4F35P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026756.1 | 4 | 2529 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP4F35P| Gene Symbol: CYP4F35P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026756.1 | 4 | 2529 | |||