siRNA Details
Gene Symbol: LINC01029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104127.1 | 2 | 371 | |||
GenBank mRNA | BC016792.1 | - | 2 | 373 | ||
BF183892.1 | - | 366 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000582805.1 | - | 2 | 366 | Chr. 18 - 77971814 - 77993722 [-] on Build GRCh38 |
