siRNA Details
| Gene Symbol: LINC01029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104127.1 | 2 | 105 | |||
| GenBank mRNA | BC016792.1 | - | 2 | 107 | ||
| BF183892.1 | - | 100 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000582805.1 | - | 2 | 100 | Chr. 18 - 77971814 - 77993722 [-] on Build GRCh38 | |

LINC01029
Genomic Map