siRNA Details
Gene Symbol: GACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120598.1 | 2 | 775 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000579368.1 | - | 2 | 775 | Chr. 18 - 8695856 - 8707621 [-] on Build GRCh38 |
