siRNA Details
| Gene Symbol: LOC105371766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135645.1 | 2 | 230 | |||
| GenBank mRNA | BX103390.1 | - | 2 | 230 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000582518.1 | - | 2 | 230 | Chr. 17 - 39026715 - 39027110 [-] on Build GRCh38 | |

LOC105371766
Genomic Map