siRNA Details
Gene Symbol: LOC105371766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135645.1 | 2 | 148 | |||
GenBank mRNA | BX103390.1 | - | 2 | 148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000582518.1 | - | 2 | 148 | Chr. 17 - 39026715 - 39027110 [-] on Build GRCh38 |
