siRNA Details
| Gene Symbol: NDUFV2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110771.1 | 3 | 502 | |||
| GenBank mRNA | BI520375.1 | - | 3 | 427 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000582375.1 | - | 3 | 427 | Chr. 18 - 9121265 - 9136645 [-] on Build GRCh38 | |

NDUFV2-AS1
Genomic Map