siRNA Details
| Gene Symbol: NDUFV2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110771.1 | 4 | 726 | |||
| NR_110772.1 | 3 | 550 | ||||
| GenBank mRNA | BI520375.1 | - | 4 | 651 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000582375.1 | - | 4 | 651 | Chr. 18 - 9121265 - 9136645 [-] on Build GRCh38 | |

NDUFV2-AS1
Genomic Map