siRNA Details
| Gene Symbol: LOC101927557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110808.1 | 3 | 404 | |||
| GenBank mRNA | AK131022.1 | - | 3 | 404 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000577189.1 | - | 3 | 404 | Chr. 17 - 57522248 - 57523597 [-] on Build GRCh38 | |

LOC101927557
Genomic Map