siRNA Details
| Gene Symbol: LOC101927666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110809.1 | 3 | 4650 | |||
| GenBank mRNA | CR936603.1 | - | 3 | 4650 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000584805.1 | - | 3 | 4650 | Chr. 17 - 58076891 - 58083204 [-] on Build GRCh38 | |

LOC101927666
Genomic Map