siRNA Details
| Gene Symbol: LINC01538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033983.1 | 3 | 1919 | |||
| GenBank mRNA | BC042493.1 | - | 3 | 1915 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588074.1 | - | 3 | 1919 | Chr. 18 - 64213082 - 64260055 [-] on Build GRCh38 | |

LINC01538
Genomic Map