siRNA Details
| Gene Symbol: LOC102724908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135636.1 | 3 | 1578 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587961.1 | - | 3 | 1578 | Chr. 19 - 28606688 - 28615229 [+] on Build GRCh38 | |

LOC102724908
Genomic Map