siRNA Details
| Gene Symbol: LINC01180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110661.1 | 1 | 1420 | |||
| GenBank mRNA | BC043289.1 | - | 1 | 1424 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589259.1 | - | 1 | 1420 | Chr. 17 - 44646364 - 44650349 [-] on Build GRCh38 | |

LINC01180
Genomic Map