siRNA Details
| Gene Symbol: LOC284263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038429.1 | - | 3 | 425 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000586386.5 | - | 3 | 423 | Chr. 18 - 45483345 - 45528306 [+] on Build GRCh38 | |

LOC284263
Genomic Map