siRNA Details
| Gene Symbol: LOC101929767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110868.1 | 1 | 163 | |||
| GenBank mRNA | AA456260.1 | - | 1 | 73 | ||
| AK311131.1 | - | 1 | 163 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000590740.2 | - | 1 | 163 | Chr. 17 - 43164183 - 43170403 [-] on Build GRCh38 | |

LOC101929767
Genomic Map