siRNA Details
| Gene Symbol: LOC101927229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110743.1 | 3 | 2110 | |||
| GenBank mRNA | AK093940.1 | - | 3 | 2110 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589125.5 | - | 3 | 2110 | Chr. 18 - 55105904 - 55124306 [-] on Build GRCh38 | |

LOC101927229
Genomic Map