siRNA Details
Gene Symbol: LINC01532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040036.1 | 5 | 897 | |||
GenBank mRNA | AK055100.1 | - | 5 | 896 | ||
BC039394.1 | - | 5 | 897 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000591143.5 | - | 5 | 897 | Chr. 19 - 29002555 - 29013955 [+] on Build GRCh38 |
