siRNA Details
| Gene Symbol: TCF4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132985.1 | 2 | 330 | |||
| GenBank mRNA | BX110727.1 | - | 2 | 330 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587660.1 | - | 2 | 330 | Chr. 18 - 55452533 - 55482940 [+] on Build GRCh38 | |

TCF4-AS1
Genomic Map