siRNA Details
| Gene Symbol: LOC100505715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038334.1 | 3 | 1745 | |||
| GenBank mRNA | BC041923.1 | - | 3 | 1759 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587128.1 | - | 3 | 1747 | Chr. 19 - 43891804 - 43901805 [-] on Build GRCh38 | |

LOC100505715
Genomic Map