siRNA Details
| Gene Symbol: DHX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525253.2 | XP_011523555.1 | 9, 10 | 1254 | ||
| Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002924.3 | 8, 9 | 744 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587125.1 | - | 8, 9 | 667 | Chr. 17 - 59962363 - 60018975 [-] on Build GRCh38 | |

DHX40
Genomic Map