siRNA Details
| Gene Symbol: LOC284294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033881.1 | 10 | 1082 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589376.1 | - | 10 | 1082 | Chr. 18 - 64104091 - 64423601 [+] on Build GRCh38 | |

LOC284294
Genomic Map