siRNA Details
| Gene Symbol: LOC101928845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134916.1 | 2 | 452 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588275.2 | - | 2 | 452 | Chr. 19 - 14333743 - 14343913 [+] on Build GRCh38 | |

LOC101928845
Genomic Map