siRNA Details
| Gene Symbol: ZNF385C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524475.2 | XP_011522777.1 | 3 | 1252 | ||
| XM_011524476.1 | XP_011522778.1 | 3 | 1203 | |||
| XM_011524477.2 | XP_011522779.1 | 3 | 1610 | |||
| XM_011524478.2 | XP_011522780.1 | 3 | 1875 | |||

ZNF385C
Genomic Map