siRNA Details
| Gene Symbol: LOC100420587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110759.1 | 10 | 2557 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000592347.5 | - | 10 | 2543 | Chr. 19 - 28435388 - 28727680 [-] on Build GRCh38 | |

LOC100420587
Genomic Map