siRNA Details
Gene Symbol: ZNF790-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040027.1 | 6 | 798 | |||
NR_040028.1 | 4 | 604 | ||||
GenBank mRNA | BM467896.1 | - | 4 | 392 | ||
BX394102.2 | - | 4 | 603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000588906.5 | - | 4 | 604 | Chr. 19 - 36797563 - 36828101 [+] on Build GRCh38 |
