siRNA Details
| Gene Symbol: LOC728485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294306.1 | NP_001281235.1 | 2 | 1496 | ||
| GenBank mRNA | AK092138.1 | - | 2 | 1475 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000590750.1 | - | 2 | 1461 | Chr. 19 - 36773153 - 36777078 [+] on Build GRCh38 | |

LOC728485
Genomic Map