siRNA Details
Gene Symbol: LOC105371795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135646.1 | 2 | 793 | |||
GenBank mRNA | AK096785.1 | - | 2 | 793 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000589796.2 | - | 2 | 793 | Chr. 17 - 45168805 - 45171485 [-] on Build GRCh38 |
