siRNA Details
| Gene Symbol: LOC105372397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134907.1 | 3 | 455 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589557.1 | - | 3 | 456 | Chr. 19 - 38596346 - 38601694 [+] on Build GRCh38 | |

LOC105372397
Genomic Map