siRNA Details
Gene Symbol: LINC01415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127787.1 | - | 2 | 3345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000587320.1 | - | 2 | 3344 | Chr. 18 - 55776727 - 55781721 [-] on Build GRCh38 |
