siRNA Details
| Gene Symbol: LINC01415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127787.1 | - | 2 | 3664 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587320.1 | - | 2 | 3663 | Chr. 18 - 55776727 - 55781721 [-] on Build GRCh38 | |

LINC01415
Genomic Map