siRNA Details
Gene Symbol: LINC01478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110792.1 | 5 | 2044 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000587877.1 | - | 5 | 2044 | Chr. 18 - 44323436 - 44531697 [-] on Build GRCh38 |
