siRNA Details
| Gene Symbol: LOC102724958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110760.1 | 4 | 1065 | |||
| GenBank mRNA | BX092781.1 | - | 4 | 636 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589623.1 | - | 2 | 176 | Chr. 19 - 28970200 - 28970874 [+] on Build GRCh38 | |

LOC102724958
Genomic Map