siRNA Details
| Gene Symbol: LINC00906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027318.2 | 2 | 1436 | |||
| GenBank mRNA | BC039341.1 | - | 2 | 1436 | ||
| BX647231.1 | - | 2 | 1434 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589921.1 | - | 2 | 1436 | Chr. 19 - 28965131 - 28969148 [+] on Build GRCh38 | |

LINC00906
Genomic Map