siRNA Details
Gene Symbol: LOC105372419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134887.1 | 2 | 592 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000586556.1 | - | 2 | 575 | Chr. 19 - 45085329 - 45090374 [-] on Build GRCh38 |
