siRNA Details
| Gene Symbol: DNAH17-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102401.1 | 1 | 56 | |||
| GenBank mRNA | BG331155.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588565.5 | - | 1 | 84 | Chr. 17 - 78484882 - 78490105 [+] on Build GRCh38 | |

DNAH17-AS1
Genomic Map