siRNA Details
| Gene Symbol: C1orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC051692.1 | - | 1 | 399 | ||
| Gene Symbol: LOC101928886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110741.1 | 4 | 1446 | |||
| GenBank mRNA | BC040925.1 | - | 4 | 1446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587218.1 | - | 4 | 1446 | Chr. 19 - 56066684 - 56078801 [+] on Build GRCh38 | |

C1orf52
Genomic Map