siRNA Details
| Gene Symbol: LOC100996351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110670.1 | 4 | 1672 | |||
| GenBank mRNA | AK123032.1 | - | 4 | 1676 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000587587.1 | - | 4 | 1672 | Chr. 19 - 3141576 - 3155175 [-] on Build GRCh38 | |

LOC100996351
Genomic Map