siRNA Details
| Gene Symbol: LOC105371899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136416.1 | 1 | 196 | |||
| NR_136417.1 | 1 | 196 | ||||
| GenBank mRNA | BM700476.1 | - | 1 | 94 | ||
| CD675441.1 | - | 1 | 37 | |||
| CD675892.1 | - | 1 | 153 | |||
| DB157978.1 | - | 1 | 196 | |||

LOC105371899
Genomic Map