siRNA Details
| Gene Symbol: LOC105371899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136416.1 | 3 | 641 | |||
| GenBank mRNA | CD173460.1 | - | 3 | 415 | ||
| CD675441.1 | - | 3 | 482 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563583.1 | - | 3 | 539 | Chr. 17 - 76962892 - 76969097 [-] on Build GRCh38 | |

LOC105371899
Genomic Map